Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A8026

Sigma-Aldrich

[Sar1, Ala8]-Angiotensin II

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H67N13O10
CAS Number:
Molecular Weight:
926.07
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](C)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Sar-Arg-Val-Tyr-Ile-His-Pro-Ala

Biochem/physiol Actions

Angiotensin II antagonist at1 and AT2 receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Pharmacology, 357, 825-825 (1976)
K Kawano et al.
European journal of pharmacology, 357(1), 33-39 (1998-10-27)
Oral administration of the angiotensin AT1 receptor antagonist 3-methyl-2,6-dimethyl-4-[[2'-(1H-tetrazol-5-yl)-1,1'-biphenyl-4-yl ]methoxy] pyridine (ME3221) inhibited the pressor response to angiotensin II at doses of 0.3-1.0 mg/kg in rats. A higher dose of ME3221 (3-10 mg/kg) was required to obtain the same inhibitory
C Caruso-Neves et al.
Biochimica et biophysica acta, 1467(1), 189-197 (2000-08-10)
Angiotensin-(1-7) (Ang-(1-7)) modulates the Na+-ATPase, but not the Na+,K+-ATPase activity present in pig kidney proximal tubules. The Na+-ATPase, insensitive to ouabain, but sensitive to furosemide, is stimulated by Ang-(1-7) (68% by 10(-9) M), in a dose-dependent manner. This effect is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service