Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A7981

Sigma-Aldrich

Anti-Aromatase antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARO, Anti-CYP XIX, Anti-CYP19A1 (Cytochrome P450, Family 19, Subfamily A, Polypeptide 1), Anti-Estrogen Synthase, Anti-P-450 Aromatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50-55 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 0.5-1 μg/mL using paraformaldehyde fixed HEK-293T cells transfected with Aromatase
western blot: 0.5-1 μg/mL using human placenta tissue lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CYP19A1(1588)

General description

The enzyme aromatase is a member of P450 cytochrome family. Aromatase catalyzes three oxidation reactions of androstenedione A that lead to aromatization of androgens and subsequent synthesis of estrogen. The enzymatic complex of aromatase is a heterocomplex of cytochrome 450 aromatase and NADPH cytochrome P450 reductase.[1][2] The tissues that produce aromatase are placenta, ovary, brain, bone, adipose tissue, skin and testis.[3] The aromatase enzyme complex is present in the smooth endoplasmic reticulum of cells of these tissues.[4] High levels of aromatase and estrogen are produced by the breast cancer cells. Selective aromatase inhibitors are developed to control the recurrence and survival of hormone-dependent breast cancer tissue[1]
Anti-Aromatase specifically recognizes human aromatase (50-55 kDa).

Application

Anti-Aromatase may be used for immunoblotting at a working concentration of 0.5-1 μg/ml using lysate of human placenta tissue. The antibody has also been used for immunoblotting using whole extracts[5] or nuclear extracts[6] of human primary trophoblasts. For immunofluorescence, a working concentration of 0.5-1 μg/ml is recommended.

Target description

Aromatase is part of the cytochrome p450 family of monooxygenases. It is involved in the production of estrogen and found localized to the endoplasmic reticulum of multiple cell types. [7]

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E A McDonald et al.
Endocrinology, 150(9), 4358-4365 (2009-06-13)
The hormone adiponectin has been shown to be important in maintaining insulin sensitivity throughout the body, whereas potential effects on the placenta have not been assessed. Pregnancy constitutes a unique physiological environment in which metabolism has a profound effect on
Ewa Monika Drzewiecka et al.
International journal of molecular sciences, 22(6) (2021-04-04)
An electromagnetic field (EMF) may affect the functions of uterine tissues. This study hypothesized that EMF changes the estrogenic activity of pig myometrium during the peri-implantation period. Tissue was collected on days 15-16 of the gestation and incubated in the
Jieyun Li et al.
Journal of animal science and biotechnology, 7, 57-57 (2016-10-07)
Heat stress is known to alter follicular dynamics and granulosa cell function and may contribute to the diminished reproductive efficiency commonly observed in mammals during the summer. Although several investigators have studied heat-induced ovarian injury, few reports have focused on
Serge Carreau et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 365(1546), 1571-1579 (2010-04-21)
In most mammalian species aromatase is encoded by a single gene (Cyp19), which contains 18 exons, nine of them being translated. In man, the presence of a biologically active aromatase and oestrogen receptors (ERalpha and ERbeta) has been reported in
Christos Symeonides et al.
Nature communications, 15(1), 6367-6367 (2024-08-08)
Male sex, early life chemical exposure and the brain aromatase enzyme have been implicated in autism spectrum disorder (ASD). In the Barwon Infant Study birth cohort (n = 1074), higher prenatal maternal bisphenol A (BPA) levels are associated with higher ASD symptoms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service