Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A7252

Supelco

Ajmaline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N2O2
CAS Number:
Molecular Weight:
326.43
EC Number:
MDL number:
UNSPSC Code:
41116107

storage temp.

2-8°C

SMILES string

CCC1C(O)N2C3CC1C4C(O)C5(CC24)[C@H]3N(C)c6ccccc56

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Indole alkaloid from Rauwolfia serpentina that is a Class Ia antiarrhythmia compound. It blocks sodium channels in skeletal and cardiac muscle and cardiac HERG potassium channels in the open state but not in the closed state. Ajmaline administered intravenously prolongs the atrioventricular (AV) conduction times in the heart and has been used diagnostically to detect AV block and other cardiac abnormalities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service