Skip to Content
MilliporeSigma
All Photos(1)

Documents

A6982

Sigma-Aldrich

HydroMatrix Peptide Cell Culture Scaffold

mixture, powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

biological source

synthetic

Quality Level

form

powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

HydroMatrix has been shown to support improved growth and migration in the proliferation of many cell types, including neural stem cells, neurons, glia, astrocytes, fibroblasts, and keratinocytes.

Components

HydroMatrix is a peptide nanofiber 3-dimensional scaffold that promotes cell growth and migration. It utilizes specific peptides that self-assemble from fluid precursors into highly crosslinked peptide hydrogels when presented with increased temperatures or ionic strengths. By inducing a rapid transformation from solution to gel, a peptide nanofiber scaffold is generated. Adjusting the concentration of the HydroMatrix solution, researchers can control the flexibility of the 3-D architecture to meet their application′s specific needs.

Caution

The lyophilized product at -20°C and is stable for many years at this temperature. The hydrogel should be stored at -70°C to maximize its active span. The 1% (w/v) stock solution should be stored -20°C and is stable for 3-6 months.

Preparation Note

This product is supplied as a lyophilized powder. A 1% (w/v) stock solution at 10 mg/mL can be prepared with addition of sterile water, resulting in a rigid gel. A 0.5% (w/v) solution will lead to a softer gel that is more suitable for cell migration and angiogenesis. Lower concentrations in the range of 0.15%-0.25% are more suitable for many cell types including neurons. In aqueous solution, HydroMatrix has a pH of 2.5 and should not be added directly to cells without prior addition of a buffer or medium. Once the hydrogel has formed, do not attempt to mix the gel any further, as this risks destroying the hydrogel integrity. Manipulation of the cells should be carefully performed to avoid disruption of the scaffold.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Stoppoloni et al.
Rheumatology international, 33(9), 2399-2403 (2012-03-28)
Osteoarthritis (OA) is one of the most common degenerative joint disease for which there is no cure. It is treated mainly with non-steroidal anti-inflammatory drugs to control the symptoms and some supplements, such as glucosamine and chondroitin sulphate in order
Amish Asthana et al.
Drug discovery today, 17(15-16), 810-817 (2012-04-10)
The three microenvironmental factors that characterize 3D cultures include: first, chemical and/or biochemical composition, second, spatial and temporal dimensions, and third, force and/or substrate physical properties. Although these factors have been studied individually, their interdependence and synergistic interactions have not
Amish Asthana et al.
Drug discovery today, 18(11-12), 533-540 (2012-12-29)
Force and substrate physical property (pliability) is one of three well established microenvironmental factors (MEFs) that may contribute to the formation of physiologically more relevant constructs (or not) for cell-based high-throughput screening (HTS) in preclinical drug discovery. In 3D cultures
Josine E G Vaes et al.
Glia, 69(3), 655-680 (2020-10-13)
Encephalopathy of prematurity (EoP) is a common cause of long-term neurodevelopmental morbidity in extreme preterm infants. Diffuse white matter injury (dWMI) is currently the most commonly observed form of EoP. Impaired maturation of oligodendrocytes (OLs) is the main underlying pathophysiological

Articles

3D cell culture overview. Learn about 2D vs 3D cell culture, advantages of 3D cell culture, and techniques available to develop 3D cell models

Hydrogela are the most widely used systems for 3D cell culture. Learn more about this technology (what are hydrogels? How to chose?)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service