Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6833

Sigma-Aldrich

Arg-Lys-Arg-Ser-Arg-Ala-Glu

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C35H67N17O11/c1-18(26(56)51-23(32(62)63)11-12-25(54)55)47-28(58)21(9-5-15-45-34(40)41)50-31(61)24(17-53)52-30(60)22(10-6-16-46-35(42)43)49-29(59)20(8-2-3-13-36)48-27(57)19(37)7-4-14-44-33(38)39/h18-24,53H,2-17,36-37H2,1H3,(H,47,58)(H,48,57)(H,49,59)(H,50,61)(H,51,56)(H,52,60)(H,54,55)(H,62,63)(H4,38,39,44)(H4,40,41,45)(H4,42,43,46)/t18-,19-,20-,21-,22-,23-,24-/m0/s1

InChI key

BVKSYBQAXBWINI-LQDRYOBXSA-N

Gene Information

human ... PRKG1(5592)

Amino Acid Sequence

Arg-Lys-Arg-Ser-Arg-Ala-Glu

Substrates

Substrate for cGMP-dependent protein kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service