Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

A6830

8-Azido-cyclic adenosine diphosphate-ribose

≥95% (HPLC), lyophilized powder

Synonym(s):

Cyclic adenosine diphosphate-ribose 8-azide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C15H20N8O13P2
CAS Number:
Molecular Weight:
582.31
UNSPSC Code:
41106305
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥95% (HPLC)

form

lyophilized powder

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H]2Nc3ncnc4[n@@H]([C@@H]5O[C@H](COP(=O)(OC[C@H]1O2)OP(O)(O)=O)[C@@H](O)[C@H]5O)c(N=[N+]=[N-])nc34

Application

Photoaffinity label

Physical form

Lyophilized powder containing approx. 50% sodium chloride.

Other Notes

Analog of cyclic ADP-ribose

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T F Walseth et al.
The Journal of biological chemistry, 268(35), 26686-26691 (1993-12-15)
We have synthesized 8-azido-cyclic ADP-ribose (8N3-cADPR) and [32P]8-azido-cyclic ADP-ribose ([32P]8N3-cADPR) in order to characterize cyclic ADP-ribose-(cADPR) binding sites in sea urchin egg homogenates. 8N3-cADPR was an antagonist of cADPR since it did not induce Ca2+ release from egg microsomes but
Preparation of cyclic ADP-ribose antagonists and caged cyclic ADP-ribose.
T F Walseth et al.
Methods in enzymology, 280, 294-305 (1997-01-01)
T F Walseth et al.
Biochimica et biophysica acta, 1178(3), 235-242 (1993-09-13)
Cyclic ADP-ribose (cADPR) is a naturally-occurring metabolite of NAD+ that is as effective as inositol trisphosphate in mobilizing intracellular Ca2+. A series of analogs modified at the 8-position of the adenine group were synthesized for the investigation of the relationship

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service