Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6791

Sigma-Aldrich

Atrial Natriuretic Peptide fragment 3-28 rat

≥97% (HPLC)

Synonym(s):

ANP, rANF 3-28

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C119H189N43O36S2
CAS Number:
Molecular Weight:
2862.17
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](CSSC[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CO)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)CNC1=O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](Cc4ccc(O)cc4)C(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)[C@@H](C)CC

Gene Information

Amino Acid Sequence

Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr [Disulfide bridge: 7-23]

Biochem/physiol Actions

Atrial natriuretic peptide (ANP) is synthesized in myoendocrine cells of the heart from which it is released into the circulation. It exerts natriuretic, diuretic, and vasodilatory effects through stimulation of guanylate cyclase-linked NPR-A receptors. It plays an important role in blood volume and blood pressure homeostasis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie A Lovshin et al.
Diabetes care, 38(1), 132-139 (2014-11-22)
GLP-1 receptor (GLP-1R) agonists induce natriuresis and reduce blood pressure (BP) through incompletely understood mechanisms. We examined the effects of acute and 21-day administration of liraglutide on plasma atrial natriuretic peptide (ANP), urinary sodium excretion, office and 24-h BP, and
Two-dimensional 1H-NMR investigation of the water conformation of the atrial natriuretic factor (ANF 101-126).
Y Theriault et al.
Biopolymers, 26(7), 1075-1086 (1987-07-01)
R Garcia et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 179(3), 396-401 (1985-07-01)
Conscious SHR and WKY rats were infused during 7 days with synthetic ANF (Arg 101-Tyr 126), 100 ng/hr/rat (35 pmol/hr/rat) by means of miniosmotic pumps. The SHR initial blood pressure of 177 +/- 5 mmHg gradually dropped to 133 +/-
I Lihrmann et al.
General and comparative endocrinology, 71(1), 55-62 (1988-07-01)
In order to investigate a possible role of atrial natriuretic factor (ANF) in the control of corticosteroid biosynthesis in amphibians, we have examined the effect of synthetic ANF (Arg 101-Tyr 126) on perifused frog interrenal slices. ANF did not affect
E L Schiffrin et al.
The Journal of clinical endocrinology and metabolism, 72(2), 484-491 (1991-02-01)
We have previously demonstrated the presence of binding sites for atrial natriuretic peptide (ANP) in human platelets. These sites have pharmacological characteristics similar to those of rat vascular smooth muscle. They are subject to regulation by circulating levels of ANP

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service