Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6668

Sigma-Aldrich

Sodium eicosyl sulfate

Synonym(s):

1-Eicosanyl sulfate, Arachidyl sulfate sodium salt, Eicosyl sodium sulfate, Eicosyl sulfate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H41O4SNa
CAS Number:
Molecular Weight:
400.59
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[Na].CCCCCCCCCCCCCCCCCCCCOS(O)(=O)=O

InChI

1S/C20H42O4S.Na.H/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-24-25(21,22)23;;/h2-20H2,1H3,(H,21,22,23);;

InChI key

DBIYGAPQOGWLPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Tyrode et al.
Nature communications, 11(1), 493-493 (2020-01-26)
Despite the importance of the hydrogen ion in a wide range of biological, chemical, and physical processes, its molecular structure in solution remains lively debated. Progress has been primarily hampered by the extreme diffuse nature of the vibrational signatures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service