Skip to Content
MilliporeSigma
All Photos(1)

Documents

A6656

Sigma-Aldrich

Anti-Human IgG (γ-chain specific)−Agarose antibody produced in goat

IgG fraction of antiserum, saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

goat

conjugate

agarose conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

saline suspension

species reactivity

human

extent of labeling

≥5 mg per mL

technique(s)

Ouchterlony double diffusion: suitable

capacity

0.6 mg/mL binding capacity

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders.
Goat anti-human IgG (γ-chain specific)-agarose antibody is specific for human IgG when tested against purified human IgA, IgG, IgM, Bence Jones kappa, and Bence Jones lambda myeloma proteins.

Immunogen

human IgG

Application

Goat anti-human IgG (γ-chain specific)-agarose antibody has been used for immunocapture assays. It can also be used for Ouchterlony double diffusion assays.
IgGs were depleted from normal human serum by incubation with 50 μl of agarose-linked rabbit anti-human antibody (A3543) for 90 minutes at 4°C.

Physical form

Suspension in 0.5 M NaCl containing preservative

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Viallard et al.
Clinical chemistry, 36(11), 2000-2003 (1990-11-01)
We report an abnormal pattern for enolase (EC 4.2.1.11) isoenzymes in the serum of a patient with squamous cell lung carcinoma. The alpha alpha-isoenzyme was present but the alpha gamma form was not detected, and near the point of application
J Raper et al.
Infection and immunity, 67(4), 1910-1916 (1999-03-20)
Natural resistance of humans to the cattle pathogen Trypanosoma brucei brucei has been attributed to the presence in human serum of nonimmune factors that lyse the parasite. Normal human serum contains two trypanosome lytic factors (TLFs). TLF1 is a 500-kDa
C Papadea et al.
Critical reviews in clinical laboratory sciences, 27(1), 27-58 (1989-01-01)
Human IgG consists of two identical heavy (H) chains and two identical light (L) chains joined by interchain disulfide bridges. Heterogeneity in the amino acid sequences of the H and L polypeptides results in at least three types of IgG
F Cataldo et al.
Minerva pediatrica, 42(12), 509-514 (1990-12-01)
IgG are the most common isotype of Ig and include four subclasses which differ from one another in the following ways: their initial amino acid sequence, their physical and chemical properties and the different serum concentrations reached with age. Every

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service