Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6561

Sigma-Aldrich

Monoclonal Anti-Biotin–Alkaline Phosphatase antibody produced in mouse

clone BN-34, purified immunoglobulin, buffered aqueous glycerol solution

Synonym(s):

Monoclonal Anti-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

alkaline phosphatase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BN-34, monoclonal

form

buffered aqueous glycerol solution

technique(s)

dot blot: 1:10,000 using human IgG and biotin-monoclonal anti-human IgG
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20 using human tonsil and biotin-monoclonal anti-human IgG
indirect ELISA: 1:30,000 using human IgG and biotin-monoclonal anti-human IgG
western blot (chemiluminescent): 1:10,000 using human IgG and biotin-monoclonal anti-human IgG

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal Anti-Biotin (mouse IgG1 isotype) is derived from the BN-34 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with biotin-keyhole limpet hemocyanin (KLH).
The antibody recognizes free biotin and biocytin using competitive ELISA. Using indirect ELISA or immuno­histo­chemical staining, the antibody also recognizes biotin conjugated to various immunoglobulins. Specificity is evaluated using biotinylated goat antibodies to human and rabbit antigens coated on multiwell plates.

Immunogen

Biotin-KLH conjugate

Application

May be substituted for avidin to decrease non-specific binding.
Monoclonal Anti-Biotin-Alkaline Phosphatase antibody produced in mouse has been used in:
  • blotting (western), Southern and dot blots)
  • immunohistology
  • in situ nucleic acids hybridization
  • enzyme linked immunosorbent assay (ELISA)
  • fluorescent activated cell-sorting (FACS)
  • electron microscopy

The antibody may be used in amplification techniques, ELISA, immunoblotting, in situ nucleic acid hybridization, flow cytometry, image analysis or confocal microscopy.
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.

Biochem/physiol Actions

Biotin is a carboxylase co-enzyme that regulates the metabolism of amino acids, glucose and fatty acids. This co-enzyme has been implicated in processes such as immune responses, cell division and fetal growth. Biotin may also be involved in the epigenetic modifications of chromatin.
The high binding affinity of biotin to egg white or bacteria-derived avidin has been exploited in the design of immunoassays and immunohistologic staining techniques. Conventional immunoassay methods are improved using monoclonal anti-biotin, which enhances the sensitivity of avidin-biotin immunoassays by adding a second layer of enzyme via the biotin specific monoclonal antibody.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David E Noga et al.
Biomacromolecules, 9(7), 2056-2062 (2008-06-26)
A polylactide copolymer with pendant benzyloxy groups has been synthesized by the copolymerization of a benzyl-ether substituted monomer with lactide. Debenzylation of the polymer to provide pendant hydroxyl groups followed by modification with succinic anhydride affords the corresponding carboxylic acid
Stereochemical engineering yields a multifunctional peptide macrocycle inhibitor of Akt2 by fine-tuning macrocycle-cell membrane interactions.
Nag, et al.
Communications Chemistry, 6, 95-95 (2023)
Spatial screening of cyclic neoglycopeptides: identification of polyvalent wheat-germ agglutinin ligands.
Valentin Wittmann et al.
Angewandte Chemie (International ed. in English), 43(7), 900-903 (2004-02-10)
Spore surface glycoproteins of Colletotrichum lindemuthianum are recognized by a monoclonal antibody which inhibits adhesion to polystyrene
Hughes H, et al.
Microbiology, 145(8), 1927-1936 (1999)
Yash M Kolambkar et al.
Tissue engineering. Part A, 20(1-2), 398-409 (2013-09-12)
Electrospun nanofiber meshes have emerged as a new generation of scaffold membranes possessing a number of features suitable for tissue regeneration. One of these features is the flexibility to modify their structure and composition to orchestrate specific cellular responses. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service