A6433
Acetyl-Ser-Asp-Lys-Pro
≥97% (HPLC)
Synonym(s):
AcSDKP, Acetyl-SDKP
About This Item
Assay
≥97% (HPLC)
form
powder
color
white
UniProt accession no.
storage temp.
−20°C
SMILES string
CC(=O)NC(CO)C(=O)NC(CC(O)=O)C(=O)NC(CCCCN)C(=O)N1CCCC1C(O)=O
InChI
1S/C20H33N5O9/c1-11(27)22-14(10-26)18(31)24-13(9-16(28)29)17(30)23-12(5-2-3-7-21)19(32)25-8-4-6-15(25)20(33)34/h12-15,26H,2-10,21H2,1H3,(H,22,27)(H,23,30)(H,24,31)(H,28,29)(H,33,34)
InChI key
HJDRXEQUFWLOGJ-UHFFFAOYSA-N
Gene Information
human ... GCG(2641)
Amino Acid Sequence
Biochem/physiol Actions
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service