Skip to Content
MilliporeSigma
All Photos(4)

Documents

A5601

Sigma-Aldrich

Anti-Arp1α/Centractin antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 45 kDa

species reactivity

mouse, human, rat

technique(s)

indirect immunofluorescence: 1:100 using mouse fibroblasts NIH3T3 cell line
microarray: suitable
western blot: 1:1,000 using rat brain cytosol extract or whole cell extract of human epidermoid carcinoma A431 cell line.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The actin-related protein Arp1α (centractin, actin-RPV), a member of the superfamily of actin-related proteins (Arp), is the major subunit of the dynactin complex, a key component of the cytoplasmic dynein motor machinery. It has been shown to localize to multiple structures within the cell, including membrane organelles, the centrosome, spindle poles, and spindle pole microtubules during mitosis and prometaphase kinetochores.

Immunogen

synthetic peptide corresponding to the mid-region of human Arp1α/centractin (amino acids 222-240). This sequence is identical in mouse and dog Arp1α/centractin, highly conserved (89% identity) in the α-centractin isoform, and not found in known actin isoforms.

Application

Anti-Arp1α/Centractin antibody produced in rabbit has been used in immunoblotting and immunofluorescence staining.

Biochem/physiol Actions

Arp1α or Centractin is an actin-related, ubiquitious protein that forms a part of the dynactin complex and is associated with centrosomes. Studies have reported that centractin interacts with spectrin and subsequently provides a link for dynactin to associate with organelles in cells.
The actin-related proteins (Arp)1α/dynactin complex regulates dyne mediated vesicle movement on microtubules as well as spindle assembly and cell division. Arp1α overexpression in cells results in aberrant spindle morphologies and cell cycle delay at prometaphase, suggesting a possible function of Arp1α/dynactin complex in progression through the prometaphase of mitosis. Expression of mutant Arp1a protein has no effect on mitotic cells, but results in microtubule disruption in interphase cells, by destabilizing the interaction between Arp1a and components of the dynactin complex and nuclear mitotic apparatus protein (NuMA).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antimitotic effect of the retinoid 4-oxo-fenretinide through inhibition of tubulin polymerization: a novel mechanism of retinoid growth-inhibitory activity
Appierto V, et al.
Molecular Cancer Therapeutics, 8(12), 3360-3368 (2009)
Regulation of spindle integrity and mitotic fidelity by BCCIP
Huhn S, et al.
Oncogene, 36(33), 4750-4750 (2017)
S W Clark et al.
Nature, 359(6392), 246-250 (1992-09-17)
Actin is one of the most ubiquitous, abundant and well-conserved proteins of eukaryotes, participating in many crucial cellular processes including the maintenance of cell shape, motility and cell division. Actins from the most divergent sources still share amino-acid identities in
S C Huhn et al.
Oncogene, 36(33), 4750-4766 (2017-04-11)
Centrosomes together with the mitotic spindle ensure the faithful distribution of chromosomes between daughter cells, and spindle orientation is a major determinant of cell fate during tissue regeneration. Spindle defects are not only an impetus of chromosome instability but are
E A Holleran et al.
The Journal of cell biology, 135(6 Pt 2), 1815-1829 (1996-12-01)
Centractin (Arp1), an actin-related protein, is a component of the dynactin complex. To investigate potential functions of the protein, we used transient transfections to overexpress centractin in mammalian cells. We observed that the overexpressed polypeptide formed filamentous structures that were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service