Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A5475

Sigma-Aldrich

ALX-1393

>98% (HPLC), powder

Synonym(s):

O-[(2-Benzyloxyphenyl-3-flurophenyl)methyl]-L-serine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22NO4F
Molecular Weight:
395.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

powder

color

white

solubility

DMSO: soluble ~20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

N[C@@H](COC(c1cccc(F)c1)c2ccccc2OCc3ccccc3)C(O)=O

InChI

1S/C23H22FNO4/c24-18-10-6-9-17(13-18)22(29-15-20(25)23(26)27)19-11-4-5-12-21(19)28-14-16-7-2-1-3-8-16/h1-13,20,22H,14-15,25H2,(H,26,27)/t20-,22?/m0/s1

InChI key

ADUSZEGHFWRTQS-AIBWNMTMSA-N

Application

ALX-1393 is a GlyT-2 glycine transporter inhibitor and GlyT2 may be a novel therapeutic target for the treatment of overactive bladder and/or bladder hypersensitive disorders such as bladder pain syndrome/interstitial cystitis.

Biochem/physiol Actions

GlyT-2 glycine transporter inhibitor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycine Transporter Type 2 (GlyT2) Inhibitor Ameliorates Bladder Overactivity and Nociceptive Behavior in Rats
Yoshikawa, S., et al.
European Urology, doi: 10-doi: 10 (2012)
Thomas M Ackermann et al.
ChemMedChem, 16(1), 199-215 (2020-08-01)
This study describes the first binding assay for glycine transporter 2 (GlyT2) following the concept of MS Binding Assays. The selective GlyT2 inhibitor Org25543 was employed as a reporter ligand and it was quantified with a highly sensitive and rapid
Birgit Nimmervoll et al.
Neuroreport, 22(10), 509-513 (2011-06-15)
To investigate whether glycine receptors influence radial migration in the neocortex, we analyzed the effect of glycine and the glycinergic antagonist strychnine, on the distribution of 5-bromo-2'deoxyuridine-labeled neurons in organotypic slice cultures from embryonic mice cortices. Application of glycine impeded
Hyo-Jin Jeong et al.
British journal of pharmacology, 161(4), 925-935 (2010-09-24)
The arachidonyl-amino acid N-arachidonyl-glycine (NAGly) is an endogenous lipid, generated within the spinal cord and producing spinally mediated analgesia via non-cannabinoid mechanisms. In this study we examined the actions of NAGly on neurons within the superficial dorsal horn, a key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service