Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5157

Sigma-Aldrich

DL-2-Amino-7-phosphonoheptanoic acid

~95%

Synonym(s):

AP-7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16NO5P
CAS Number:
Molecular Weight:
225.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

~95%

SMILES string

NC(CCCCCP(O)(O)=O)C(O)=O

InChI

1S/C7H16NO5P/c8-6(7(9)10)4-2-1-3-5-14(11,12)13/h6H,1-5,8H2,(H,9,10)(H2,11,12,13)

InChI key

MYDMWESTDPJANS-UHFFFAOYSA-N

Biochem/physiol Actions

Selective NMDA glutamate receptor antagonist; anticonvulsant.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M K Bazil et al.
The American journal of physiology, 260(1 Pt 2), H267-H275 (1991-01-01)
These studies investigated the role of spinal N-methyl-D-aspartic acid (NMDA) receptors in the mediation of cardiovascular responses evoked by L-glutamate (L-Glu) stimulation of the rostral ventrolateral medulla (RVM). Microinjections of L-Glu into the RVM of urethan-anesthetized rats increased mean arterial
M Benveniste et al.
British journal of pharmacology, 104(1), 207-221 (1991-09-01)
1. The kinetics of action of 17 structurally related NMDA receptor competitive antagonists were measured under voltage clamp in mouse hippocampal neurones. Analysis of the response to rapid changes in antagonist concentration during constant application of agonist was used to
Ariandra G Sartim et al.
Behavioural pharmacology, 30(1), 59-66 (2018-10-10)
The transient receptor potential vanilloid 1 (TRPV1) can modulate stress-related behaviours, thus representing an interesting target for new antidepressant drugs. TRPV1 can trigger glutamate release and nitric oxide synthesis in the brain, mechanisms also involved in the neurobiology of depression.
Luiz F S Souza-Pinto et al.
Pharmacology, biochemistry, and behavior, 87(2), 250-257 (2007-06-01)
It is well established that the most persistent sign of withdrawal from chronic benzodiazepine use in humans is anxiety. In contrast to other types of drugs of abuse, the emergence of this anxiety does not seem to be linked directly
Leandro José Bertoglio et al.
Behavioural pharmacology, 17(7), 589-596 (2006-10-06)
The involvement of the dorsolateral periaqueductal gray in the regulation of fear-related behaviors such as escape and freezing is well established. It is still a matter of investigation, however, whether this midbrain area may have a relevant role in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service