Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5058

Sigma-Aldrich

Ala-Pro-Arg-Leu-Arg-Phe-Tyr

≥97% (HPLC)

Synonym(s):

α-Bag Cell Peptide 1-7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H67N13O9
CAS Number:
Molecular Weight:
922.08
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ~70%

storage temp.

−20°C

InChI

1S/C44H67N13O9/c1-25(2)22-32(54-37(60)31(13-8-20-51-44(48)49)53-40(63)35-14-9-21-57(35)41(64)26(3)45)38(61)52-30(12-7-19-50-43(46)47)36(59)55-33(23-27-10-5-4-6-11-27)39(62)56-34(42(65)66)24-28-15-17-29(58)18-16-28/h4-6,10-11,15-18,25-26,30-35,58H,7-9,12-14,19-24,45H2,1-3H3,(H,52,61)(H,53,63)(H,54,60)(H,55,59)(H,56,62)(H,65,66)(H4,46,47,50)(H4,48,49,51)

InChI key

NXTLZAZOGADWPA-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R S Redman et al.
Brain research. Molecular brain research, 8(2), 99-103 (1990-07-01)
alpha-Bag cell peptide (alpha-BCP), one of several secreted peptides encoded in the precursor to the egg-laying hormone (proELH) of the neurosecretory bag cells of Aplysia, has been variously reported to have autoexcitatory or autoinhibitory effects on the cells which secrete
B S Rothman et al.
Proceedings of the National Academy of Sciences of the United States of America, 80(18), 5753-5757 (1983-09-01)
A discharge of impulse activity in a group of neuroendocrine cells, the bag cells, produces several types of prolonged responses in various identified neurons of the abdominal ganglion of Aplysia. Two excitatory responses are almost certainly mediated by egg-laying hormone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service