Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A5048

Sigma-Aldrich

Adenosine 5′-diphosphate–Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500

form

saline suspension

extent of labeling

1-3 μmol per mL

matrix

4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

ribose hydroxyls

matrix spacer

11 atoms (adipic acid dihydrazide)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Adenosine 5′-diphosphate has been used in the research of platelet integrin α(IIb)β3, which is crucial for platelet aggregation. It has been determined that the interaction between Adenosine 5′-diphosphate and the receptor P2Y12 is needed for the maintenance of integrin α(IIb)β3 activation.

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C R Tinsley et al.
Infection and immunity, 61(9), 3703-3710 (1993-09-01)
The important human pathogens Neisseria meningitidis and Neisseria gonorrhoeae accumulate phosphate in the form of polyphosphate (A. Noegel and E. C. Gotschlich, J. Exp. Med. 157:2049-2060, 1983), and the localization of more than half of this long-chain polymer on the
T Kamae et al.
Journal of thrombosis and haemostasis : JTH, 4(6), 1379-1387 (2006-05-19)
Platelet integrin alpha(IIb)beta3 plays a crucial role in platelet aggregation, and the affinity of alpha(IIb)beta3 for fibrinogen is dynamically regulated. Employing modified ligand-binding assays, we analyzed the mechanism by which alpha(IIb)beta3 maintains its high-affinity state. Washed platelets adjusted to 50
Knut Fälker et al.
Thrombosis and haemostasis, 92(1), 114-123 (2004-06-24)
Stimulating human platelets with thrombin induces the activation of the extracellular signal-regulated kinase 2 (ERK2). We demonstrate that this effect is highly dependent on ADP secretion and P2Y12 receptor signalling. AR-C69931MX (10 microM), a specific antagonist of the Gi-coupled P2Y12
Francesca Campus et al.
The Journal of biological chemistry, 280(26), 24386-24395 (2005-05-03)
Binding of thrombopoietin (TPO) to the cMpl receptor on human platelets potentiates aggregation induced by a number of agonists, including ADP. In this work, we found that TPO was able to restore ADP-induced platelet aggregation upon blockade of the G(q)-coupled

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service