Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4815

Sigma-Aldrich

Acetyl-pepstatin

≥95% (HPLC)

Synonym(s):

Pepstatin, acetyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H57N5O9
CAS Number:
Molecular Weight:
643.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)CC(NC(=O)C(C)NC(=O)CC(O)C(CC(C)C)NC(=O)C(NC(=O)C(NC(C)=O)C(C)C)C(C)C)C(O)CC(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Richards et al.
FEBS letters, 247(1), 113-117 (1989-04-10)
Inhibitory constants (Ki) between 5 and 35 nM were derived (under different conditions of pH and ionic strength) for the interaction of HIV-1 proteinase with acetyl-pepstatin and H-261, two characteristic inhibitors of aspartic proteinases. Thus this enzyme, essential for replication

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service