Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4803

Sigma-Aldrich

Adenosine 5′-triphosphate γ-(1-[2-nitrophenyl]ethyl) ester sodium salt

≥95%

Synonym(s):

“Caged” ATP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21N6Na2O15P3
CAS Number:
Molecular Weight:
700.29
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥95%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(OP([O-])(=O)OP([O-])(=O)OP(O)(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23)c4ccccc4[N+]([O-])=O

InChI

1S/C18H23N6O15P3.2Na/c1-9(10-4-2-3-5-11(10)24(27)28)37-41(31,32)39-42(33,34)38-40(29,30)35-6-12-14(25)15(26)18(36-12)23-8-22-13-16(19)20-7-21-17(13)23;;/h2-5,7-9,12,14-15,18,25-26H,6H2,1H3,(H,29,30)(H,31,32)(H,33,34)(H2,19,20,21);;/q;2*+1/p-2/t9?,12-,14-,15-,18-;;/m1../s1

InChI key

DWGOHIKVVKFGGU-DZHDPEGHSA-L

Application

Adenosine 5′-triphosphate γ-(1-[2-nitrophenyl]ethyl) ester (“Caged” ATP) is used as a photolyzing substrate of luciferase-mediated firefly bioluminescence and other ATP-dependent photolytic processes. Caged ATP has also been used to study the dynamics of ATP-driven linear molecular motors such as myosin Va.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuyuki Shiroguchi et al.
PLoS biology, 9(4), e1001031-e1001031 (2011-05-03)
Myosins are ATP-driven linear molecular motors that work as cellular force generators, transporters, and force sensors. These functions are driven by large-scale nucleotide-dependent conformational changes, termed "strokes"; the "power stroke" is the force-generating swinging of the myosin light chain-binding "neck"
Christian Völlmecke et al.
The FEBS journal, 276(21), 6172-6186 (2009-09-29)
The mechanism of ATP hydrolysis of a shortened variant of the heavy metal-translocating P-type ATPase CopB of Sulfolobus solfataricus was studied. The catalytic fragment, named CopB-B, comprises the nucleotide binding and phosphorylation domains. We demonstrated stoichiometric high-affinity binding of one
Takeshi Kageyama et al.
Photochemistry and photobiology, 87(3), 653-658 (2011-01-07)
The reaction process of firefly bioluminescence was studied by photolyzing caged-ATP to adenosine triphosphate (ATP) within 100 ms. The intensity of luminescence increases markedly to reach a maximum within 1 s, maintains almost the same intensity up to 5 s
J A McCray et al.
Proceedings of the National Academy of Sciences of the United States of America, 77(12), 7237-7241 (1980-12-01)
2-Nitrobenzyl derivatives have been used for several years as photolabile protecting groups in synthetic organic chemistry. Recently, P3-1-(2-nitro phenylethyladenosine 5'-triphosphate "caged ATP" was synthesized and its photolysis was shown to generate ATP in situ. This and related reactions have great
Savas Tasoglu et al.
Advanced materials (Deerfield Beach, Fla.), 27(26), 3901-3908 (2015-06-11)
A simple, yet powerful magnetic-levitation-based device is reported for real-time, label-free separation, as well as high-resolution monitoring of cell populations based on their unique magnetic and density signatures. This method allows a wide variety of cellular processes to be studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service