Skip to Content
MilliporeSigma
All Photos(1)

Documents

A4602

Sigma-Aldrich

Arg-Arg-Ala-pSer-Pro-Val-Ala

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H58N13O12P
Molecular Weight:
835.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

color

white

solubility

H2O: 1 mg/mL

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@H](COP(O)(O)=O)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C31H58N13O12P/c1-15(2)22(27(49)40-17(4)29(51)52)43-26(48)21-10-7-13-44(21)28(50)20(14-56-57(53,54)55)42-23(45)16(3)39-25(47)19(9-6-12-38-31(35)36)41-24(46)18(32)8-5-11-37-30(33)34/h15-22H,5-14,32H2,1-4H3,(H,39,47)(H,40,49)(H,41,46)(H,42,45)(H,43,48)(H,51,52)(H4,33,34,37)(H4,35,36,38)(H2,53,54,55)/t16-,17-,18-,19-,20-,21-,22-/m0/s1

InChI key

SYLMVFSDXGIMAW-CPDXTSBQSA-N

Amino Acid Sequence

Arg-Arg-Ala-pSer-Pro-Val-Ala

Biochem/physiol Actions

The phosphoseryl peptide RRApSPVA can provide a sensitive evaluation of the majority of acid and alkaline phosphatases, while being refractory to protein phosphatases.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiwen Liu et al.
JCI insight, 5(24) (2020-12-18)
Sepsis is the leading cause of acute kidney injury (AKI). However, the pathogenesis of septic AKI remains largely unclear. Here, we demonstrate a significant decrease of microRNA-376b (miR-376b) in renal tubular cells in mice with septic AKI. Urinary miR-376b in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service