Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4535

Sigma-Aldrich

5-(2-Aminoethylamino)-1-naphthalenesulfonic acid sodium salt

suitable for fluorescence

Synonym(s):

1,5-EDANS, N-(Aminoethyl)-5-naphthylamine-1-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C12H13N2O3SNa
CAS Number:
Molecular Weight:
288.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

suitability

suitable for fluorescence

SMILES string

[Na+].NCCNc1cccc2c(cccc12)S([O-])(=O)=O

InChI

1S/C12H14N2O3S.Na/c13-7-8-14-11-5-1-4-10-9(11)3-2-6-12(10)18(15,16)17;/h1-6,14H,7-8,13H2,(H,15,16,17);/q;+1/p-1

InChI key

HGWRACRQRUQQGH-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Schilling et al.
Biological chemistry, 399(9), 997-1007 (2018-06-09)
In humans, three different trypsin-isoenzymes have been described. Of these, trypsin-3 appears to be functionally different from the others. In order to systematically study the specificity of the trypsin-isoenzymes, we utilized proteome-derived peptide libraries and quantitative proteomics. We found similar
Claudia Seidl et al.
Current Alzheimer research, 14(3), 317-326 (2016-10-28)
Alzheimer´s disease, a progressive and degenerative disorder of the brain, is the most common cause of dementia among the elderly. To face its multifactorial nature, the use of single compounds that can simultaneously modulate different targets involved in the neurodegenerative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service