Skip to Content
MilliporeSigma
All Photos(1)

Documents

A4316

Sigma-Aldrich

Anantin from Streptomyces coerulescens

≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C90H111N21O24
CAS Number:
Molecular Weight:
1870.97
MDL number:
UNSPSC Code:
12352200

Assay

≥90% (HPLC)

storage temp.

−20°C

Amino Acid Sequence

Cyclo(Gly-Phe-Ile-Gly-Trp-Gly-Asn-β-Asp)-Ile-Phe-Gly-His-Tyr-Ser-Gly-Asp-Phe

Biochem/physiol Actions

Competitive antagonist at atrial natriuretic peptide NPR-A receptors.

Other Notes

A cyclic peptide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D F Wyss et al.
The Journal of antibiotics, 44(2), 172-180 (1991-02-01)
Anantin, a naturally occurring peptide from Streptomyces coerulescens, binds competitively to the receptor of atrial natriuretic factor (ANF) from bovine adrenal cortex (Kd = 0.6 microM) and acts as ANF antagonist. Protein chemical data and FAB-MS have identified anantin to
Rouwayda El-Ayoubi et al.
British journal of pharmacology, 145(1), 50-56 (2005-02-09)
We have previously shown that acute intravenous injections of moxonidine and clonidine increase plasma atrial natriuretic peptide (ANP), a vasodilator, diuretic and natriuretic hormone. We hypothesized that moxonidine stimulates the release of ANP, which would act on its renal receptors
Sandra L Hrometz et al.
Toxicology letters, 206(2), 234-237 (2011-08-11)
MDMA (3,4-methylenedioxymethamphetamine) induces thermogenesis in a mitochondrial uncoupling protein 3-dependent manner. There is evidence that this hyperthermia is mediated in part by the lipolytic release of free fatty acids, that subsequently activate uncoupling protein 3 in skeletal muscle mitochondria. We
S Nachshon et al.
The American journal of physiology, 268(3 Pt 1), E428-E432 (1995-03-01)
Atrial natriuretic peptide (ANP) is a hormone-secreted predominantly by atrial myocytes. ANP exerts many of its actions via activation of the particulate guanylyl cyclase receptor ANPR-A and the formation of guanosine 3',5'-cyclic monophosphate (cGMP), which serves as a second messenger
Martin Kerr et al.
Current biology : CB, 14(16), 1468-1474 (2004-08-25)
Every living cell must detect, and respond appropriately to, external signals. The functions of intracellular second messengers, such as guanosine 3'5'-cyclic monophosphate (cGMP), adenosine 3'5'-cyclic monophosphate (cAMP), and intracellular calcium, are thus intensively studied. However, artifact-free manipulation of these messengers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service