Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4183

Sigma-Aldrich

Apolipoprotein B amide fragment 3358-3372

≥97% (HPLC)

Synonym(s):

Thr-Arg-Leu-Thr-Arg-Lys-Arg-Gly-Leu-Lys-Leu-Ala-Thr-Ala-Leu-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C74H141N27O18
CAS Number:
Molecular Weight:
1697.08
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(C)C)NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)[C@@H](C)O)[C@@H](C)O)[C@@H](C)O)C(N)=O

Gene Information

human ... APOB(338)

Biochem/physiol Actions

Anti-proliferative peptide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Cardin et al.
Biochemical and biophysical research communications, 154(2), 741-745 (1988-07-29)
Apolipoproteins B and E of the human plasma lipoproteins are known inhibitors of lymphocyte proliferation. In this report, two synthetic peptide amides, apoB3358-3372 and apoE141-155, showed a dose-dependent inhibition of both the murine mixed lymphocyte culture reaction and the anti-T3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service