Skip to Content
Merck
All Photos(1)

Key Documents

A4146

Sigma-Aldrich

Alizarin Yellow R sodium salt

Synonym(s):

Mordant orange 1, Mordant yellow 3R

Sign Into View Organizational & Contract Pricing


About This Item

Colour Index Number:
14030
EC Number:
UNSPSC Code:
12171500

Pricing and availability is not currently available.

composition

Dye content, ~65%

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA031369HPA043359HPA038600
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... CPNE3(8895)

Gene Information

human ... CPNE5(57699)

Gene Information

human ... CPNE7(27132)

Gene Information

human ... CPNE8(144402)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service