Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A4095

Sigma-Aldrich

Anti-phospho-ATF-2 (pThr69,71) antibody, Mouse monoclonal

clone ATF-22P, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-phospho-ATF-2 (pThr69,71)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ATF-22P, monoclonal

form

buffered aqueous solution

mol wt

antigen 60-70 kDa (triplet)

species reactivity

frog, mouse, rat, chicken, human

concentration

0.5-1.0 mg/mL

technique(s)

immunocytochemistry: suitable
indirect ELISA: suitable
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr69/pThr71)

Gene Information

human ... ATF2(1386)
mouse ... Atf2(11909)
rat ... Atf2(81647)

Looking for similar products? Visit Product Comparison Guide

General description

ATF2 (Activating Transcription Factor 2) is a member of the leucine zipper family of transcription factors. It binds to CRE domains and stimulates CRE-dependent transcription.
Mouse monoclonal anti-phospho-ATF-2 (pThr69,71) reacts specifically with ATF2 phosphorylated at both threonine 69 and 71, and does not detect the monophosphorylated and the non-phosphorylated ATF2 molecule. The epitope recognized by the antibody resides in the N-terminal region of ATF2 (aa 66-79, pT69,71). This epitope is identical in ATF2 of human, rat, mouse, chicken and frog. The antibody cross-reacts with human, rat and mouse ATF2.

Immunogen

synthetic peptide corresponding to the N-terminal amino acids 66-79 (pT69,71) of human ATF2. This sequence is identical in ATF2 from human, rat, mouse, chicken, and frog.

Application

Mouse monoclonal anti-phospho-ATF-2 (pThr69,71) antibody can be used for immunocytochemistry, indirect ELISA, microarray, and western blot.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Purified from culture supernatant of hybridoma cells grown in a bioreactor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service