Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3771

Sigma-Aldrich

8-(6-Aminohexyl)aminoadenosine 5′-monophosphate lithium salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N7O7P
CAS Number:
Molecular Weight:
461.41
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

[Li].NCCCCCCNc1nc2c(N)ncnc2n1C3OC(COP(O)(O)=O)C(O)C3O

InChI

1S/C16H28N7O7P.Li.H/c17-5-3-1-2-4-6-19-16-22-10-13(18)20-8-21-14(10)23(16)15-12(25)11(24)9(30-15)7-29-31(26,27)28;;/h8-9,11-12,15,24-25H,1-7,17H2,(H,19,22)(H2,18,20,21)(H2,26,27,28);;

InChI key

AZVYMTYCFZUNFT-UHFFFAOYSA-N

Application

Ligand for preparation of affinity chromatography medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guilford, H., et al.
The Biochemical Journal, 139, 44-44 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service