Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3609

Sigma-Aldrich

AC-55541

≥98% (HPLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20BrN5O3
Molecular Weight:
518.36
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: >5 mg/mL

storage temp.

2-8°C

SMILES string

C\C(=N/NC(=O)C(NC(=O)c1ccccc1)C2=NNC(=O)c3ccccc23)c4cccc(Br)c4

InChI

1S/C25H20BrN5O3/c1-15(17-10-7-11-18(26)14-17)28-31-25(34)22(27-23(32)16-8-3-2-4-9-16)21-19-12-5-6-13-20(19)24(33)30-29-21/h2-14,22H,1H3,(H,27,32)(H,30,33)(H,31,34)/b28-15+

InChI key

UCUHFWIFSHROPY-RWPZCVJISA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung-Hsing Chao et al.
Journal of biomedical science, 24(1), 85-85 (2017-11-17)
This study investigated whether lipopolysaccharide (LPS) increase protease-activated receptor-2 (PAR-2) expression and enhance the association between PAR-2 expression and chemokine production in human vascular endothelial cells (ECs). The morphology of ECs was observed through microphotography in cultured human umbilical vein
Fabio Rabelo Melo et al.
Cell death & disease, 10(9), 659-659 (2019-09-12)
It is well established that mast cell accumulation accompanies most malignancies. However, the knowledge of how mast cells functionally impact on tumors is still rudimentary. Here we addressed this issue and show that mast cells have anti-proliferative activity on melanoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service