Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A3418

Sigma-Aldrich

Anti-α1-Acid Glycoprotein antibody produced in goat

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen 40.8 kDa (sedimentation velocity and diffusion studies)

contains

15 mM sodium azide

species reactivity

human

technique(s)

quantitative precipitin assay: 1.5 mg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ORM1(5004)

General description

α-1 acid glycoprotein is a protein at 43kDa secreted mainly from the liver during acute phase reaction. Its expression is induced by cytokines and corticosteroids.
Alpha-1-acid glycoprotein (AGP) is also known as orosomucoid (ORM). It is a 41-43 kDa glycoprotein. It is one of the important acute phase proteins found in humans, rats, mice and other species. This immunosuppressive glycoprotein is composed of two isoforms, such as ORM1 and ORM2.
Goat Anti-α1-Acid Glycoprotein antibody is immunospecific for human α1-acid glycoprotein by immunoelectrophoresis versus human serum and α1-acid glycoprotein.

Immunogen

purified human α1-acid glycoprotein

Application

Goat Anti-α1-Acid Glycoprotein antibody can be used for quantitative precipitin assays at 1.5mg/ml.

Biochem/physiol Actions

Abberations of orosomucoid 1 (ORM1) is linked with the pharmacokinetics of telmisartan. This plasma protein participates in the binding and transport of several drugs.

Physical form

Supplied as a liquid containing 15mM sodium azide as preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphism of ORM1 Is Associated with the Pharmacokinetics of Telmisartan
Chen WQ, et al.
Testing, 8(8) (2013)
Effect of alpha-1-acid glycoprotein binding on pharmacokinetics and pharmacodynamics
Huang Z, et al.
Current Drug Metabolism, 14(2), 226-238 (2013)
Alpha-1-acid glycoprotein
Fournier T, et al.
Biochimica et Biophysica Acta, 1482(1-2), 157-171 (2000)
Elisabetta Razzuoli et al.
Veterinary research communications, 47(3), 1615-1627 (2023-04-11)
Canine Soft Tissue Sarcoma (STS) cell line A-72 has been largely employed for antiviral and antiproliferative studies. However, there are few information on their characteristics. Our aim was to evaluate A-72 expression level of genes and proteins involved in the
Kiwamu Nakamura et al.
Cytokine, 73(1), 8-15 (2015-02-18)
Orosomucoid (ORM, composed of two isoforms, ORM1 and ORM2) has been described as an inducer of M2 macrophages, which are cells that decrease host antibacterial innate immunities. However, it is unknown which phenotypes of M2 macrophages are induced by ORM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service