Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A2941

Sigma-Aldrich

Arachidonoyl chloride

~95% (capillary GC, NMR)

Synonym(s):

5,8,11,14-Eicosatetraenoic acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31ClO
CAS Number:
Molecular Weight:
322.91
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~95% (capillary GC, NMR)

density

0.92 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(Cl)=O

InChI

1S/C20H31ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h6-7,9-10,12-13,15-16H,2-5,8,11,14,17-19H2,1H3/b7-6-,10-9-,13-12-,16-15-

InChI key

NTBLHFFUSJRJQO-DOFZRALJSA-N

Packaging

Sealed ampule

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Smyrniotis et al.
Biochemistry, 53(27), 4407-4419 (2014-06-04)
5-Lipoxygenase (5-LOX) reacts with arachidonic acid (AA) to first generate 5(S)-hydroperoxy-6(E),8(Z),11(Z),14(Z)-eicosatetraenoic acid [5(S)-HpETE] and then an epoxide from 5(S)-HpETE to form leukotriene A4, from a single polyunsaturated fatty acid. This work investigates the kinetic mechanism of these two processes and
Kui Yang et al.
Analytical and bioanalytical chemistry, 407(17), 5199-5210 (2015-03-31)
Diglycerides play a central role in lipid metabolism and signaling in mammalian cells. Although diacylglycerol molecular species comprise the majority of cellular diglycerides that are commonly measured using a variety of approaches, identification of extremely low abundance vinyl ether diglycerides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service