Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2903

Sigma-Aldrich

Acyl Coenzyme A Oxidase from Candida sp.

lyophilized powder, 10-30 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

10-30 units/mg protein

storage temp.

−20°C

Unit Definition

One unit will form 1.0 μmole of H2O2 and hexadecenoyl-CoA from palmitoyl-CoA per min at pH 8.0 at 30 °C in a peroxidase coupled system.

Physical form

Contains sucrose as stabilizer; protein determined by biuret

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuhua Sun et al.
American journal of physiology. Gastrointestinal and liver physiology, 302(5), G548-G557 (2011-12-17)
The development of alcohol-induced fatty liver is associated with a reduction of white adipose tissue (WAT). Peroxisome proliferator-activated receptor (PPAR)-γ prominently distributes in the WAT and plays a crucial role in maintaining adiposity. The present study investigated the effects of
Nana Mikami et al.
Journal of food science, 77(6), H114-H120 (2012-05-16)
n-3 Polyunsaturated fatty acids (PUFAs) such as eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA), and taurine are functional compounds abundantly present in seafoods. In this study, we examined the combined effects of EPA- and DHA-rich fish oil and taurine on
Hiromu Ohnogi et al.
Bioscience, biotechnology, and biochemistry, 76(5), 928-932 (2012-06-29)
Angelica keiskei is a traditional herb peculiar to Japan and abundantly contains vitamins, dietary fiber and such polyphenols as chalcone. We investigated in the present study the effect of A. keiskei on insulin resistance and hypertriglyceridemia in fructose-drinking rats as
Meiping Tian et al.
Toxicology, 296(1-3), 48-55 (2012-03-20)
Perfluorooctanoic acid (PFOA) is one of the most commonly used perfluorinated compounds. Being a persistent environmental pollutant, it can accumulate in human tissues via various exposure routes. PFOA may interfere in a toxic fashion on the immune system, liver, development
Eriko Misawa et al.
Journal of agricultural and food chemistry, 60(11), 2799-2806 (2012-02-23)
We investigated the effects of the oral administration of lophenol (Lo) and cycloartanol (Cy), two kinds of antidiabetic phytosterol isolated from Aloe vera , on glucose and lipid metabolism in Zucker diabetic fatty (ZDF) rats. We demonstrated that the administrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service