Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2734

Sigma-Aldrich

α5IA

≥98% (HPLC)

Synonym(s):

1,2,4-Triazolo[3,4-a]phthalazine, 3-(5-methyl-3-isoxazolyl)-6-[(1-methyl-1H-1,2,3-triazol-4-yl)methoxy]-, 3-(5-Methylisoxazol-3-yl)-6-(1-methyl-1,2,3-triazol-4-yl)methoxy-1,2,4-triazolo[3,4-a]phthalazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N8O2
CAS Number:
Molecular Weight:
362.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white to light yellow

solubility

DMSO: >2 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

SMILES string

Cc1cc(no1)-c2nnc3c4ccccc4c(OCc5cn(C)nn5)nn23

InChI

1S/C17H14N8O2/c1-10-7-14(22-27-10)16-20-19-15-12-5-3-4-6-13(12)17(21-25(15)16)26-9-11-8-24(2)23-18-11/h3-8H,9H2,1-2H3

InChI key

NZMJFRXKGUCYNP-UHFFFAOYSA-N

Biochem/physiol Actions

α5IA is a selective inverse agonist for Α5 subtype of GABAA receptor with a higher intrinsic activity at the A5 subtype than other drugs. α5IA enhances cognition in laboratory animals without being proconvulsant or anxiogenic. α5IA acts as an "alcohol antagonist," markedly reducing the amnesic effect of alcohol and partially attenuating sedation, similar to Ro15-4513, but without the anxiogenic effect. α5IA offers promise as a tool to study cognitive disorders, memory function, and treatment of alcohol-related disorders.
α5IA is a selective inverse agonist for a5 subtype of GABAA receptor.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service