Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2349

Sigma-Aldrich

Adenosine-3′,5′-cyclic monophosphorothioate acetoxymethyl ester, Sp-isomer

>97% (HPLC), solid

Synonym(s):

Sp-cAMPS-AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N5O7SP
Molecular Weight:
417.33
MDL number:
UNSPSC Code:
41106305

Assay

>97% (HPLC)

form

solid

packaging

vial of 1.0 μmol

storage temp.

−20°C

Biochem/physiol Actions

Highly membrane-permeable precursor of the phosphodiesterase-resistant protein kinase A activator sp-cAMPS (Prod. No. A-166).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service