Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2224

Sigma-Aldrich

Adenosine 3′,5′-cyclic monophosphate acetoxymethyl ester

>97% (HPLC)

Synonym(s):

cAMP AM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N5O8P
CAS Number:
Molecular Weight:
401.27
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

>97% (HPLC)

form

solid

packaging

pkg of 1 μmol

storage temp.

−20°C

SMILES string

CC(=O)OCOP1(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O1)n3cnc4c(N)ncnc34

InChI

1S/C13H16N5O8P/c1-6(19)22-5-24-27(21)23-2-7-10(26-27)9(20)13(25-7)18-4-17-8-11(14)15-3-16-12(8)18/h3-4,7,9-10,13,20H,2,5H2,1H3,(H2,14,15,16)/t7-,9-,10-,13-,27?/m1/s1

InChI key

USKCKKDBQFHVEK-OQEAWBJFSA-N

Biochem/physiol Actions

Membrane-permeable precursor of the second messenger cyclic AMP

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Schultz et al.
The Journal of biological chemistry, 268(9), 6316-6322 (1993-03-25)
Acetoxymethyl esters of alkyl or aryl phosphates can be prepared by reacting their trialkylammonium or silver salts with acetoxymethyl bromide. Because acetoxymethyl esters are rapidly cleaved intracellularly, they facilitate the delivery of organophosphates into the cytoplasm without puncturing or disruption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service