Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2108

Sigma-Aldrich

AURORA B, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AIK2, AIM1, ARK2, AURKB, AurB, IPL1, STK12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

162-219 nmol/min·mg

mol wt

~68 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AURKB(9212)

Biochem/physiol Actions

AURORA B is a member of the Aurora kinase family that associates with microtubules during chromosome movement and segregation. AURORA B localizes to the microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. AURORA B inhibits the microtubule depolymerizing activity of mitotic centromere-associated kinesin (MCAK) by phosphorylating MCAK on Ser92.This phosphorylation also regulates MCAK translocalization from kinetochores to the centromere. AURORA B has been identified as a target for the development of new anticancer agents since inhibition of AURORA B gives rise to the more pronounced antiproliferative phenotype.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul D Andrews et al.
Developmental cell, 6(2), 253-268 (2004-02-13)
Chromosome orientation and alignment within the mitotic spindle requires the Aurora B protein kinase and the mitotic centromere-associated kinesin (MCAK). Here, we report the regulation of MCAK by Aurora B. Aurora B inhibited MCAK's microtubule depolymerizing activity in vitro, and
Katie B Shannon et al.
Current biology : CB, 12(13), R458-R460 (2002-07-18)
Aurora B family kinases play an essential role in chromosome segregation and cytokinesis. Recent work suggests that the kinase activity is required for bipolar chromosome orientation, kinetochore assembly, spindle checkpoint and microtubule dynamics. Aurora B also has additional functions in

Questions

1–2 of 2 Questions  
  1. What is an appropriate substrate to use as a positive control when testing product A2108, AURORA B, active, GST tagged human?

    1 answer
    1. The recommended product to use as a positive control substrate for product A2108, AURORA B, active, GST tagged human, is product SRP5205, MBP, GST tagged human.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service