Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A202

Sigma-Aldrich

N6-2-(4-Aminophenyl)ethyladenosine

>95%

Synonym(s):

APNEA, Adenosine, N6-2-(4-aminophenyl)ethyl, N-[2-(4-Aminophenyl)ethyl]-adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N6O4
CAS Number:
Molecular Weight:
386.41
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

>95%

form

solid

solubility

methanol: slightly soluble 0.2 mg/mL
0.1 M HCl: 14 mg/mL
DMSO: soluble
H2O: insoluble

storage temp.

2-8°C

SMILES string

Nc1ccc(CCNc2ncnc3n(cnc23)[C@@H]4O[C@H](CO)[C@@H](O)[C@H]4O)cc1

InChI

1S/C18H22N6O4/c19-11-3-1-10(2-4-11)5-6-20-16-13-17(22-8-21-16)24(9-23-13)18-15(27)14(26)12(7-25)28-18/h1-4,8-9,12,14-15,18,25-27H,5-7,19H2,(H,20,21,22)/t12-,14-,15-,18-/m1/s1

InChI key

XTPOZVLRZZIEBW-SCFUHWHPSA-N

Gene Information

Biochem/physiol Actions

Potent, non-selective A3 adenosine receptor agonist.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Collis et al.
Trends in pharmacological sciences, 14(10), 360-366 (1993-10-01)
The numerous and widespread effects of adenosine provide both an opportunity for the development of novel therapeutic agents acting via adenosine receptors and the challenge of achieving selectivity of action. The feasibility of achieving selectivity is enhanced if receptor subtypes
Adenosine A3 receptors: two into one won't go.
A M Carruthers et al.
Trends in pharmacological sciences, 14(8), 290-291 (1993-08-01)
J R Fozard et al.
British journal of pharmacology, 109(1), 3-5 (1993-05-01)
The cardiovascular effects of N6-2-(4-aminophenyl)ethyladenosine (APNEA), which when radiolabelled with 125I shows high affinity for the newly described adenosine A3 receptor, have been investigated in the angiotensin II-supported circulation of the pithed rat. APNEA induces hypotensive responses which are unaffected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service