Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A2002

Sigma-Aldrich

Adenosine 5′-monophosphate monohydrate

from yeast, ≥99%

Synonym(s):

5′-AMP, A-5′-P, AMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O7P · H2O
CAS Number:
Molecular Weight:
365.24
Beilstein:
54612
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

biological source

yeast

Assay

≥99%

form

powder

solubility

H2O: soluble (with addition of mild alkali)

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@H](N(C=N2)C3=C2C(N)=NC=N3)O[C@@H]1COP(O)(O)=O.O

InChI

1S/C10H14N5O7P.H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(22-10)1-21-23(18,19)20;/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,20);1H2/t4-,6-,7-,10-;/m1./s1

InChI key

ZOEFQKVADUBYKV-MCDZGGTQSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanzhong Xu et al.
Biochemical and biophysical research communications, 451(2), 184-189 (2014-07-30)
The melanocortin receptor 4 (MC4R) plays a major role in body weight regulation and its agonist MTII has been widely used to study the role of MC4Rs in energy expenditure promotion and feeding reduction. Unexpectedly, we observed that intraperitoneal (i.p.)
Yi-Fang Yang et al.
Cancer letters, 485, 27-37 (2020-05-18)
The tumor microenvironment (TME) and metabolic reprogramming have been implicated in cancer development and progression. However, the link between TME, metabolism, and cancer progression in lung cancer is unclear. In the present study, we identified IMPAD1 from the conditioned medium
Song Chen et al.
Experimental eye research, 127, 26-36 (2014-06-01)
CD73 is intensively involved in the regulation of immune responses through the conversion of pro-inflammatory ATP to immunosuppressive adenosine. Herein, we clarified whether cells in the retina express CD73 and participate in the regulation of inflammatory eye diseases such as
Yuanyuan Yang et al.
International journal of oncology, 47(1), 244-252 (2015-05-13)
Interaction between multi-functional mesenchymal stroma/stem cells (MSC) and human tumor cells involves the exchange of biological material via extracellular vesicles including exosomes. Protein analysis of MSC-derived exosomes demonstrated the presence of MMP-2 and MSC-specific markers including CD90 and ecto-5'-nucleotidase (CD73).
Lizhen Qiao et al.
Journal of chromatography. A, 1360, 240-247 (2014-08-19)
Glucaminium-based ionic liquids are a new class of recently developed ionic liquids and prepared by functionalizing the amine group of N-methyl-d-glucamine, which renders them good hydrophilicity due to the presence of the glucose structure and charged quaternary ammonium group. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service