Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A199

Sigma-Aldrich

Aminoguanidine hemisulfate salt

Synonym(s):

Aminoguanidine hemisulfate salt, Guanylhydrazine hemisulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH6N4 · 1/2H2SO4
CAS Number:
Molecular Weight:
123.12
EC Number:
MDL number:
UNSPSC Code:
12352204
Pricing and availability is not currently available.

SMILES string

NNC(N)=N.NNC(N)=N.OS(O)(=O)=O

InChI

1S/2CH6N4.H2O4S/c2*2-1(3)5-4;1-5(2,3)4/h2*4H2,(H4,2,3,5);(H2,1,2,3,4)

InChI key

VKGQPUZNCZPZKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits both constitutive and inducible nitric oxide synthetase

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service