Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

A1911

Sigma-Aldrich

AMPSO sodium salt

Synonym(s):

3-([1,1-Dimethyl-2-hydroxyethyl]amino)-2-hydroxypropanesulfonic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$689.00

$689.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$689.00

About This Item

Linear Formula:
C7H16NO5SNa
CAS Number:
Molecular Weight:
249.26
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

$689.00


Please contact Customer Service for Availability

Request a Bulk Order

form

crystalline powder

Quality Level

useful pH range

8.3-9.7

pKa (25 °C)

9.0 (Free acid)

solubility

water: 0.33 g/mL, clear to slightly hazy, colorless

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].CC(C)(CO)NCC(O)CS([O-])(=O)=O

InChI

1S/C7H17NO5S.Na/c1-7(2,5-9)8-3-6(10)4-14(11,12)13;/h6,8-10H,3-5H2,1-2H3,(H,11,12,13);/q;+1/p-1

InChI key

UVGYCHPTWHWEPZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service