Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1599

Sigma-Aldrich

[des-Arg1]-Angiotensin III

≥95% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H54N8O8
Molecular Weight:
774.91
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

form

lyophilized powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](N)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanna Andersson et al.
Journal of medicinal chemistry, 54(11), 3779-3792 (2011-04-12)
Macrocyclic analogues of angiotensin IV (Ang IV, Val(1)-Tyr(2)-Ile(3)-His(4)-Pro(5)-Phe(6)) targeting the insulin-regulated aminopeptidase (IRAP) have been designed, synthesized, and evaluated biologically. Replacement of His(4)-Pro(5)-Phe(6) by a 2-(aminomethyl)phenylacetic acid (AMPAA) moiety and of Val(1) and Ile(3) by amino acids bearing olefinic side
Aneta Lukaszuk et al.
Journal of medicinal chemistry, 51(7), 2291-2296 (2008-04-05)
Angiotensin IV, a metabolite of angiotensin II, inhibits the enzyme insulin regulated aminopeptidase or IRAP and also, although with lower potency, aminopeptidase-N (AP-N). When both beta (2)-homo amino acid- and beta (3)-homo amino acid substitutions were used, allowed the identification
Hanna Andersson et al.
Bioorganic & medicinal chemistry, 16(14), 6924-6935 (2008-06-17)
Analogues of the hexapeptide angiotensin IV (Ang IV, Val(1)-Tyr(2)-Ile(3)-His(4)-Pro(5)-Phe(6)) encompassing a 4-hydroxydiphenylmethane scaffold replacing Tyr(2) and a phenylacetic or benzoic acid moiety replacing His(4)-Pro(5)-Phe(6) have been synthesized and evaluated in biological assays. The analogues inhibited the proteolytic activity of cystinyl
K Hermann et al.
Journal of neurochemistry, 52(3), 863-868 (1989-03-01)
The degradation pattern and rate of [Ile5]-Angiotensin (Ang) I, II, and III were studied in neuron-enriched and glia-enriched cells in primary cultures from rat brain. Metabolites were separated by HPLC, and their identities were evaluated by comparison of their retention
Aneta Lukaszuk et al.
Journal of medicinal chemistry, 52(18), 5612-5618 (2009-09-18)
The histidine residue in angiotensin IV was replaced by various conformationally constrained amino acids. The substitution of the His(4)-Pro(5) dipeptide sequence by the constrained Trp analogue Aia-Gly, in combination with beta(2)hVal substitution at the N-terminus, provided a new stable analogue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service