Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1314

Sigma-Aldrich

3′-N-Acetylneuraminyl-3-fucosyllactose sodium salt

Synonym(s):

α-Neu-5Ac-(2→3)-β-D-Gal-(1→4)-(α-Fuc-[1→3])-Glc, SFL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C29H48NO23Na
CAS Number:
Molecular Weight:
801.67
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC1OC(OC(C(O)C=O)C(OC2OC(CO)C(O)C(OC3(CC(O)C(NC(C)=O)C(O3)C(O)C(O)CO)C(O)=O)C2O)C(O)CO)C(O)C(O)C1O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Shen et al.
Analytical biochemistry, 279(1), 37-45 (2000-02-23)
Oligosaccharides in human milk inhibit enteric pathogens in vitro and in vivo. Neutral milk oligosaccharides vary among individuals and over the course of lactation. To study such variation in the acidic milk oligosaccharides, a sensitive, convenient, quantitative method is needed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service