A1284
(S)-trans-2-Amino-4-(2-aminoethoxy)-3-butenoic acid hydrochloride
≥95% (titration)
Synonym(s):
L-α-(2-Aminoethoxyvinyl)glycine hydrochloride, AVG-HCl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H12N2O3 · HCl
CAS Number:
Molecular Weight:
196.63
Beilstein:
6715149
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥95% (titration)
≥98% (HPLC)
storage temp.
−20°C
SMILES string
Cl[H].NCCO\C=C\[C@H](N)C(O)=O
InChI
1S/C6H12N2O3.ClH/c7-2-4-11-3-1-5(8)6(9)10;/h1,3,5H,2,4,7-8H2,(H,9,10);1H/b3-1+;/t5-;/m0./s1
InChI key
ZDCPLYVEFATMJF-BTIOQYSDSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Inhibition of ethylene biosynthesis delays ripening of fruits in plants treated with AVG.
replaced by
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qian-Yi Tao et al.
Yi chuan = Hereditas, 32(6), 632-638 (2010-06-23)
Sex determination in plants has various mechanism; however, a single gene locus controlling unisexual expression is unique in the Cucurbitaceae plants, particularly cucumber (Cucumis sativus L.) and melon (Cucumis melo L.). In this study, with quantitative RT-PCR method, two sets
Lee-Chung Lin et al.
The Journal of biological chemistry, 285(43), 33445-33456 (2010-08-05)
Ethylene is a gaseous hormone important for adaptation and survival in plants. To further understand the signaling and regulatory network of ethylene, we used a phenotype-based screening strategy to identify chemical compounds interfering with the ethylene response in Arabidopsis thaliana.
Soo Cheon Chae et al.
TheScientificWorldJournal, 2012, 859381-859381 (2012-11-06)
Shoot organogenesis and plant regeneration in Sinningia speciosa were improved using ethylene inhibitors. The leaf explants were cultured on initial shoot regeneration media (MS media with BAP at 2 mg/L + NAA at 0.1 mg/L) supplemented with different concentrations of aminoethoxyvinylglycine (AVG), cobalt chloride
Chenshuo Chang et al.
Journal of plant physiology, 167(14), 1152-1156 (2010-07-08)
Ethylene is an important plant gas hormone, and the amino acid Glu is emerging as a messenger molecule in plants. To evaluate the role of ethylene and Glu in seed germination and radicle growth under salt stress, effects of 1-aminocyclopropane-1-carboxylic
Pratik H Lodha et al.
Protein science : a publication of the Protein Society, 19(3), 383-391 (2009-12-17)
Cystathionine beta-lyase (CBL) catalyzes the hydrolysis of L-cystathionine (L-Cth) to produce L-homocysteine, pyruvate, and ammonia. A series of active-site mutants of Escherichia coli CBL (eCBL) was constructed to investigate the roles of residues R58, R59, D116, W340, and R372 in
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service