Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A1021

Sigma-Aldrich

2′-Azido-2′-deoxyuridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N5O5
CAS Number:
Molecular Weight:
269.21
Beilstein:
4204828
EC Number:
MDL number:
UNSPSC Code:
12352204

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H]([C@H](N=[N+]=[N-])[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H11N5O5/c10-13-12-6-7(17)4(3-15)19-8(6)14-2-1-5(16)11-9(14)18/h1-2,4,6-8,15,17H,3H2,(H,11,16,18)/t4-,6-,7-,8-/m1/s1

InChI key

MRUKYOQQKHNMFI-XVFCMESISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service