Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0881

Sigma-Aldrich

p-Anisidine

crystalline

Synonym(s):

4-Aminoanisole, 4-Methoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NH2
CAS Number:
Molecular Weight:
123.15
Beilstein:
471556
EC Number:
MDL number:
UNSPSC Code:
12352200

form

crystalline

autoignition temp.

959 °F

color

dark gray to brown

bp

240-243 °C (lit.)

mp

56-59 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(N)cc1

InChI

1S/C7H9NO/c1-9-7-4-2-6(8)3-5-7/h2-5H,8H2,1H3

InChI key

BHAAPTBBJKJZER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Carc. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

251.6 °F - closed cup

Flash Point(C)

122 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Héberger et al.
Lipids, 34(1), 83-92 (1999-04-03)
Decomposition of hydroperoxides in sunflower oil under strictly oxygen-free conditions was followed by measuring peroxide values against time, absorbance values at 232 and 268 nm, para-anisidine values, and by quantitative analyses of volatile products using various additives. The results were
Francesco Caponio et al.
Journal of food science, 74(9), C701-C706 (2010-05-25)
An experimental investigation was carried out to evaluate the influence of processing and storage time on the lipidic fraction of taralli. The data obtained pointed out that the kneading phase caused a significant increase of the oxidized triacylglycerols and triacylglycerol
C Meier et al.
Carcinogenesis, 12(9), 1633-1640 (1991-09-01)
We report here on the conformational analysis of C8-arylamine nucleoside and nucleotide adducts of the borderline carcinogens 4-methylaniline and 4-methoxyaniline. The non-phosphorylated adducts show anti conformation of the glycosidic link, while the corresponding 5'-phosphorylated adducts have a syn conformation. All
James S Harvey et al.
Mutagenesis, 20(1), 51-56 (2005-01-27)
The pH 6.7 Syrian hamster embryo (SHE) cell morphological transformation assay is a short-term in vitro test that has been used to predict rodent carcinogenicity. Previous reports have indicated that the SHE assay has an overall concordance of approximately 80%
Yujiro Hayashi et al.
Nature protocols, 2(1), 113-118 (2007-04-03)
This protocol describes a procedure for the synthesis of syn-beta-amino alpha-substituted aldehydes, versatile intermediates in synthetic organic chemistry, via asymmetric, direct, one-pot, three-component, cross-Mannich reaction of two different aldehydes. The reaction consists of two steps; one is the formation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service