Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0847

Sigma-Aldrich

Adenylyl Cyclase Toxin from Bordetella pertussis

≥70% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, ≥50 units/mg protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106305

recombinant

expressed in E. coli

Quality Level

Assay

≥70% (SDS-PAGE)

form

lyophilized powder

specific activity

≥50 units/mg protein

mol wt

177 kDa by amino acid sequence

storage temp.

2-8°C

Gene Information

human ... CAMP(820)
mouse ... CAMP(12796)
rat ... CAMP(316010)

Biochem/physiol Actions

Adenylate cyclase toxin from Bordetella pertussis belongs to the family of pore-forming toxins. It converts host ATP to cyclic AMP (cAMP). May be used to affect intracellular cAMP levels in mammalian cells.

Unit Definition

One unit will produce 1.0 μmol cAMP from ATP per minute at pH 8.0 at 30 °C

Reconstitution

When reconstituted with 50 μL of water, the protein is in 0.05 M Tris, 8 M urea, 0.002 M EDTA, pH 8.0.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service