Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0811

Sigma-Aldrich

Anthranilyl-His-Lys-Ala-Arg-Val-Leu-p-nitro-Phe-Glu-Ala-Nle-Ser amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H100N20O17
CAS Number:
Molecular Weight:
1433.61
MDL number:
UNSPSC Code:
12352204

composition

Peptide content, ~70%

storage temp.

−20°C

SMILES string

CCCCC(NC(=O)C(C)NC(=O)C(CCC(O)=O)NC(=O)C(Cc1ccc(cc1)N(=O)=O)NC(=O)C(CC(C)C)NC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)C(C)NC(=O)C(CCCCN)NC(=O)C(Cc2cnc[nH]2)NC(=O)c3ccccc3N)C(C)C)C(=O)NC(CO)C(N)=O

Substrates

HIV protease substrate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dunn, B.M., et al.
Abstract - 12th American Peptide Symposium (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service