Skip to Content
MilliporeSigma
All Photos(1)

Documents

A0806

Sigma-Aldrich

Acetyl-Ser-Gln-Asn-Tyr-Pro-Val-Val amide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H58N10O12
CAS Number:
Molecular Weight:
846.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(NC(=O)C1CCCN1C(=O)C(Cc2ccc(O)cc2)NC(=O)C(CC(N)=O)NC(=O)C(CCC(N)=O)NC(=O)C(CO)NC(C)=O)C(C)C)C(N)=O

Substrates

HIV-1 protease substrate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Moore et al.
Biochemical and biophysical research communications, 159(2), 420-425 (1989-03-15)
Oligopeptides containing the consensus retroviral protease cleavage sequence Ser/Thr-X-Y-Tyr/Phe-Pro are substrates for purified recombinant HIV-1 protease with Km's in the millimolar range. The minimum sequence containing the consensus pentapeptide which serves as a good substrate is a heptapeptide spanning the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service