Skip to Content
Merck
All Photos(1)

Key Documents

A0590

Millipore

Amies transport medium with charcoal

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

shelf life

limited shelf life, expiry date on the label

final pH

7.2±0.2 (25 °C)

application(s)

microbiology

Application

Used for transportation and preservation of microbiological specimens.

Components

Components (g/L)
Sodium chloride 3.00
Potassium chloride 0.20
Calcium chloride 0.10
Magnesium chloride 0.10
Monopotassium phosphate 0.20
Disodium phosphate 1.15
Sodium thioglycollate 1.00
Charcoal 10.00

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1 - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service