Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A0519

Sigma-Aldrich

p-Anisaldehyde

Synonym(s):

4-Methoxybenzaldehyde, Aubépine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CHO
CAS Number:
Molecular Weight:
136.15
Beilstein:
471382
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

4.7 (vs air)

Assay

≥98% (GC)

refractive index

n20/D 1.573 (lit.)

bp

248 °C (lit.)

mp

−1 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccc(OC)cc1

InChI

1S/C8H8O2/c1-10-8-4-2-7(6-9)3-5-8/h2-6H,1H3

InChI key

ZRSNZINYAWTAHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by A88107-ALDRICH | p-Anisaldehyde 98%

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service