Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

95620

Sigma-Aldrich

p-Xylenolphthalein

for microscopy

Synonym(s):

2′,5′,2″,5″-Tetramethylphenolphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22O4
CAS Number:
Molecular Weight:
374.43
Beilstein:
346799
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

for microscopy

form

powder

SMILES string

Cc1cc(c(C)cc1O)C2(OC(=O)c3ccccc23)c4cc(C)c(O)cc4C

InChI

1S/C24H22O4/c1-13-11-21(25)15(3)9-19(13)24(20-10-16(4)22(26)12-14(20)2)18-8-6-5-7-17(18)23(27)28-24/h5-12,25-26H,1-4H3

InChI key

PXCIPOXPHMTCIL-UHFFFAOYSA-N

Application

p-Xylenolphthalein is related to phenolphthalein which is used as a pH change indicator.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service