Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92326

Sigma-Aldrich

8-Hydroxyquinoline β-D-galactopyranoside

≥98.0% (HPLC)

Synonym(s):

8-Quinolinyl-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17NO6
CAS Number:
Molecular Weight:
307.30
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥98.0% (HPLC)

form

powder

optical activity

[α]/D -86.0±4.0°, c = 1 in methanol

impurities

≤10% water

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2cccc3cccnc23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C15H17NO6/c17-7-10-12(18)13(19)14(20)15(22-10)21-9-5-1-3-8-4-2-6-16-11(8)9/h1-6,10,12-15,17-20H,7H2/t10-,12+,13+,14-,15-/m1/s1

InChI key

BWMXDESAZVPVGR-BGNCJLHMSA-N

Biochem/physiol Actions

Chromogenic substrate for β-D-galactosidase

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection of specific bacterial enzymes by high contrast metal chelate formation. Part I. 8-Hydroxyquinoline-β-D-glucoside, an alternative to aesculin in the differentiation of members of the family Enterobacteriaceae
James, A.L. and Yeoman, P.
Zentralblatt Fur Bakteriologie, Mikrobiologie Und Hygiene. 1. Abt. Originale A, Medizinische Mikrobiologie, Infektionskrankheiten Und Parasitologie = International Journal of Microbiology and Hygiene. A, Medical Microbiology, Infectious..., 188-193 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service