Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91257

Sigma-Aldrich

MTSEA Biotin-X

solubility: in DMSO, soluble

Synonym(s):

2-((6-((Biotinoyl)amino)hexanoyl)amino)ethyl methanethiosulfonate, MTSEA-Biotincap

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H34N4O5S3
Molecular Weight:
494.69
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

crystals

shelf life

limited shelf life, expiry date on the label

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

CS(=O)(=O)SCCNC(=O)CCCCCNC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12

InChI

1S/C19H34N4O5S3/c1-31(27,28)30-12-11-21-17(25)8-3-2-6-10-20-16(24)9-5-4-7-15-18-14(13-29-15)22-19(26)23-18/h14-15,18H,2-13H2,1H3,(H,20,24)(H,21,25)(H2,22,23,26)/t14-,15-,18-/m0/s1

InChI key

PXMZUZLKVZKKHK-MPGHIAIKSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Thiol-reactive reagent for labeling proteins at cysteine residues to study structure and function.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service